Assay Details
Gene Symbol: Tbcel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014089.1 | NP_001014111.1 | 4 - 5 | 670 | 100 | |
XM_006242859.2 | XP_006242921.1 | 4 - 5 | 3317 | 100 | ||
XM_008766134.1 | XP_008764356.1 | 5 - 6 | 3280 | 100 | ||
GenBank mRNA | BC087163.1 | - | 4 - 5 | 670 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
