Assay Details
Gene Symbol: Cers4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107117.1 | NP_001100587.1 | 8 - 9 | 792 | 76 | |
XM_006248804.2 | XP_006248866.1 | 7 - 8 | 908 | 76 | ||
XM_006248805.2 | XP_006248867.1 | 7 - 8 | 810 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
