Assay Details
Gene Symbol: F11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047848.1 | NP_001041313.1 | 6 - 7 | 718 | 74 | |
XM_006253144.2 | XP_006253206.1 | 7 - 8 | 1020 | 74 | ||
XM_006253145.1 | XP_006253207.1 | 5 - 6 | 629 | 74 | ||
GenBank mRNA | AY387072.1 | - | 6 - 7 | 718 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
