Assay Details
Gene Symbol: Ccdc134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024355.1 | NP_001019526.1 | 4 - 5 | 699 | 133 | |
XM_006242113.2 | XP_006242175.1 | 5 - 6 | 612 | 133 | ||
GenBank mRNA | BC088206.1 | - | 4 - 5 | 699 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
