Assay Details
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244855.1 | NP_001231784.1 | 1 - 2 | 176 | 113 | |
GenBank mRNA | CV126932.1 | - | 1 - 2 | 176 | 113 | |
FQ217036.1 | - | 1 - 2 | 160 | 113 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001244855.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
