Assay Details
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244855.1 | NP_001231784.1 | 4 - 5 | 513 | 70 | |
XM_006242100.2 | XP_006242162.1 | 4 - 5 | 1953 | 70 | ||
XM_006242101.2 | XP_006242163.1 | 4 - 5 | 802 | 70 | ||
GenBank mRNA | BC082056.1 | - | 4 - 5 | 491 | 70 | |
CV126932.1 | - | 4 - 5 | 513 | 70 | ||
FQ217036.1 | - | 4 - 5 | 497 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
