Assay Details
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244855.1 | NP_001231784.1 | 5 - 6 | 697 | 81 | |
XM_006242100.2 | XP_006242162.1 | 5 - 6 | 2137 | 81 | ||
XM_006242101.2 | XP_006242163.1 | 5 - 6 | 986 | 81 | ||
GenBank mRNA | BC082056.1 | - | 5 - 6 | 675 | 81 | |
DY317647.1 | - | 1 - 2 | 169 | 81 | ||
FQ217036.1 | - | 5 - 6 | 681 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
