Assay Details
Gene Symbol: Maf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014085.1 | NP_001014107.1 | 6 - 7 | 975 | 105 | |
XM_006241811.1 | XP_006241873.1 | 6 - 7 | 1050 | 105 | ||
GenBank mRNA | BC083712.1 | - | 6 - 7 | 975 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
