Assay Details
Gene Symbol: Naaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010967.2 | NP_001010967.1 | 4 - 5 | 622 | 82 | |
GenBank mRNA | AB162193.1 | - | 4 - 5 | 620 | 82 | |
BC105771.1 | - | 4 - 5 | 622 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
