Assay Details
Gene Symbol: Kctd20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256148.1 | XP_006256210.1 | 3 - 4 | 531 | 73 | |
XM_006256149.1 | XP_006256211.1 | 3 - 4 | 522 | 73 | ||
XM_006256151.2 | XP_006256213.1 | 2 - 3 | 443 | 73 | ||
XM_006256152.2 | XP_006256214.1 | 2 - 3 | 404 | 73 | ||
XM_228042.6 | XP_228042.4 | 2 - 3 | 740 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
