Assay Details
Gene Symbol: Srpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025726.1 | NP_001020897.1 | 10 - 11 | 1067 | 76 | |
XM_008772734.1 | XP_008770956.1 | 10 - 11 | 1101 | 76 | ||
GenBank mRNA | BC099089.1 | - | 10 - 11 | 1067 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
