Assay Details
Gene Symbol: Srpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025726.1 | NP_001020897.1 | 11 - 12 | 1487 | 127 | |
GenBank mRNA | BC099089.1 | - | 11 - 12 | 1487 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
