Assay Details
Gene Symbol: Rrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025740.1 | NP_001020911.1 | 6 - 7 | 718 | 131 | |
GenBank mRNA | BC099082.1 | - | 6 - 7 | 718 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
