Assay Details
Gene Symbol: Ndufaf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008318.1 | NP_001008319.1 | 3 - 4 | 301 | 107 | |
XM_006239627.2 | XP_006239689.1 | 3 - 4 | 408 | 107 | ||
GenBank mRNA | BC083810.1 | - | 3 - 4 | 301 | 107 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
