Assay Details
Gene Symbol: Ndufaf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008318.1 | NP_001008319.1 | 6 - 7 | 683 | 100 | |
XM_006239627.2 | XP_006239689.1 | 6 - 7 | 790 | 100 | ||
GenBank mRNA | BC083810.1 | - | 6 - 7 | 683 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
