Assay Details
Gene Symbol: Dld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199385.2 | NP_955417.1 | 10 - 11 | 1136 | 117 | |
XM_008764590.1 | XP_008762812.1 | 10 - 11 | 1153 | 117 | ||
GenBank mRNA | BC062069.1 | - | 10 - 11 | 1140 | 117 | |
FM046215.1 | - | 2 - 3 | 306 | 117 | ||
FQ215299.1 | - | 10 - 11 | 1143 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
