Assay Details
Gene Symbol: Acp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135562.1 | NP_001129034.1 | 2 - 3 | 187 | 96 | |
XM_008764572.1 | XP_008762794.1 | 2 - 3 | 187 | 96 | ||
GenBank mRNA | BC158763.1 | - | 2 - 3 | 187 | 96 | |
BC161990.1 | - | 2 - 3 | 133 | 96 | ||
FQ215147.1 | - | 2 - 3 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
