Assay Details
Gene Symbol: Mfsd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729615.3 | XP_002729661.1 | 2 - 3 | 536 | 67 | |
XM_006239869.2 | XP_006239931.1 | 2 - 3 | 536 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
