Assay Details
Gene Symbol: Mfsd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729615.3 | XP_002729661.1 | 7 - 8 | 1082 | 63 | |
XM_006239869.2 | XP_006239931.1 | 7 - 8 | 1109 | 63 | ||
XM_006239870.2 | XP_006239932.1 | 6 - 7 | 983 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
