Assay Details
Gene Symbol: Creb3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007093.1 | NP_001007094.1 | 6 - 7 | 730 | 96 | |
XM_006232627.2 | XP_006232689.1 | 7 - 8 | 788 | 96 | ||
XM_008761147.1 | XP_008759369.1 | 6 - 7 | 885 | 96 | ||
XM_008761148.1 | XP_008759370.1 | 6 - 7 | 960 | 96 | ||
GenBank mRNA | AY750875.1 | - | 6 - 7 | 730 | 96 | |
AY750876.1 | - | 6 - 7 | 724 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
