Assay Details
Gene Symbol: Etfdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198742.2 | NP_942037.2 | 11 - 12 | 1639 | 69 | |
XM_006232507.1 | XP_006232569.1 | 10 - 11 | 1492 | 69 | ||
XM_008761084.1 | XP_008759306.1 | 12 - 13 | 1865 | 69 | ||
GenBank mRNA | AY365022.1 | - | 11 - 12 | 1465 | 69 | |
BC081890.1 | - | 11 - 12 | 1639 | 69 | ||
FQ211230.1 | - | 11 - 12 | 1696 | 69 | ||
FQ218772.1 | - | 13 - 14 | 2006 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
