Assay Details
Gene Symbol: Npm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203340.1 | NP_976085.1 | 2 - 3 | 146 | 69 | |
XM_006252261.2 | XP_006252323.1 | 3 - 4 | 956 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
