Assay Details
Gene Symbol: Mycl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191763.1 | NP_001178692.1 | 1 - 2 | 496 | 79 | |
XM_006238793.2 | XP_006238855.1 | 2 - 3 | 612 | 79 | ||
XM_006238795.2 | XP_006238857.1 | 2 - 3 | 703 | 79 | ||
XM_008764002.1 | XP_008762224.1 | 2 - 3 | 649 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
