Assay Details
Gene Symbol: Edar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191899.1 | NP_001178828.1 | 1 - 2 | 52 | 59 | |
XM_006256381.2 | XP_006256443.1 | 2 - 3 | 345 | 59 | ||
GenBank mRNA | FQ211923.1 | - | 2 - 3 | 280 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
