Assay Details
Gene Symbol: Blmh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034163.1 | NP_001029335.1 | 7 - 8 | 972 | 62 | |
GenBank mRNA | BC128701.1 | - | 7 - 8 | 857 | 62 | |
CB583895.1 | - | 1 - 2 | 108 | 62 | ||
CB768995.1 | - | 1 - 2 | 62 | |||
D87336.1 | - | 7 - 8 | 875 | 62 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
