Assay Details
Gene Symbol: Map3k10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191621.3 | NP_001178550.1 | 1 - 2 | 1016 | 66 | |
XM_006228576.2 | XP_006228638.1 | 1 - 2 | 1225 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
