Assay Details
Gene Symbol: Slc5a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107007.1 | NP_001100477.1 | 11 - 12 | 1245 | 75 | |
XM_008767820.1 | XP_008766042.1 | 11 - 12 | 1445 | 75 | ||
GenBank mRNA | BC161867.1 | - | 11 - 12 | 1438 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
