Assay Details
Gene Symbol: Slc9c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769699.1 | XP_008767921.1 | 22 - 23 | 2873 | 77 | |
XM_008769700.1 | XP_008767922.1 | 21 - 22 | 2806 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
