Assay Details
Gene Symbol: Slc9c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769699.1 | XP_008767921.1 | 25 - 26 | 3367 | 85 | |
XM_008769700.1 | XP_008767922.1 | 24 - 25 | 3300 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
