Assay Details
Gene Symbol: Slc9c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769699.1 | XP_008767921.1 | 9 - 10 | 1161 | 113 | |
XM_008769700.1 | XP_008767922.1 | 8 - 9 | 1094 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
