Assay Details
Gene Symbol: Nars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034921.1 | NP_001030093.1 | 7 - 8 | 1061 | 72 | |
GenBank mRNA | BC088466.1 | - | 218 | 72 | ||
BC104691.1 | - | 7 - 8 | 1061 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
