Assay Details
Gene Symbol: Nars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034921.1 | NP_001030093.1 | 9 - 10 | 1195 | 94 | |
GenBank mRNA | BC088466.1 | - | 352 | 94 | ||
BC104691.1 | - | 9 - 10 | 1195 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
