Assay Details
Gene Symbol: Ndufa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199495.2 | NP_955789.2 | 1 - 2 | 136 | 159 | |
XM_008767385.1 | XP_008765607.1 | 1 - 2 | 148 | 159 | ||
GenBank mRNA | CB610197.1 | - | 1 - 2 | 136 | 159 | |
FQ209525.1 | - | 1 - 2 | 138 | 159 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_199495.2, XM_008767385.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
