Assay Details
Gene Symbol: Lancl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053723.1 | NP_446175.1 | 2 - 3 | 349 | 66 | |
GenBank mRNA | AJ131111.1 | - | 2 - 3 | 349 | 66 | |
AJ295233.1 | - | 2 - 3 | 357 | 66 | ||
BC085811.1 | - | 2 - 3 | 327 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
