Assay Details
Gene Symbol: Hus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251468.1 | XP_006251530.1 | 8 | 1601 | 155 | |
XM_006251469.2 | XP_006251531.1 | 8 | 1805 | 155 | ||
XM_006251470.2 | XP_006251532.1 | 7 | 1477 | 155 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
