Assay Details
Gene Symbol: Bcs1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007666.1 | NP_001007667.1 | 4 - 5 | 958 | 151 | |
GenBank mRNA | BC083660.1 | - | 4 - 5 | 958 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
