Assay Details
Gene Symbol: Actr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009268.1 | NP_001009268.1 | 7 - 8 | 940 | 148 | |
GenBank mRNA | BC088442.1 | - | 7 - 8 | 940 | 148 | |
FQ212547.1 | - | 7 - 8 | 939 | 148 | ||
FQ212814.1 | - | 8 - 9 | 904 | 148 | ||
FQ219838.1 | - | 7 - 8 | 906 | 148 | ||
FQ225424.1 | - | 7 - 8 | 911 | 148 | ||
FQ226083.1 | - | 7 - 8 | 886 | 148 | ||
FQ226422.1 | - | 7 - 8 | 889 | 148 | ||
FQ227142.1 | - | 7 - 8 | 889 | 148 | ||
FQ227830.1 | - | 7 - 8 | 889 | 148 | ||
FQ230292.1 | - | 3 - 4 | 432 | 148 | ||
FQ230874.1 | - | 7 - 8 | 925 | 148 | ||
FQ231828.1 | - | 7 - 8 | 891 | 148 | ||
FQ231874.1 | - | 7 - 8 | 924 | 148 | ||
FQ234230.1 | - | 7 - 8 | 891 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
