Assay Details
Gene Symbol: Mrpl38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009369.2 | NP_001009369.2 | 4 - 5 | 606 | 112 | |
XM_008768372.1 | XP_008766594.1 | 4 - 5 | 1394 | 112 | ||
GenBank mRNA | BC087096.1 | - | 4 - 5 | 603 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
