Assay Details
Gene Symbol: Cdh19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009448.1 | NP_001009448.1 | 1 - 2 | 201 | 133 | |
XM_006249653.2 | XP_006249715.1 | 2 - 3 | 839 | 133 | ||
GenBank mRNA | AB121032.1 | - | 1 - 2 | 201 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
