Assay Details
Gene Symbol: Riok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009687.1 | NP_001009687.1 | 3 - 4 | 328 | 144 | |
GenBank mRNA | BC088297.1 | - | 3 - 4 | 328 | 144 | |
FQ232861.1 | - | 3 - 4 | 363 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
