Assay Details
Gene Symbol: Sh3glb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009692.1 | NP_001009692.1 | 11 - 12 | 961 | 144 | |
XM_006233816.2 | XP_006233878.1 | 12 - 13 | 1088 | 144 | ||
XM_006233818.2 | XP_006233880.1 | 11 - 12 | 937 | 144 | ||
XM_006233819.2 | XP_006233881.1 | 10 - 11 | 925 | 144 | ||
XM_006233822.2 | XP_006233884.1 | 9 - 10 | 862 | 144 | ||
XM_008761661.1 | XP_008759883.1 | 11 - 12 | 1076 | 144 | ||
GenBank mRNA | BC087652.1 | - | 11 - 12 | 961 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
