Assay Details
Gene Symbol: Asphd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009716.1 | NP_001009716.1 | 3 - 4 | 1474 | 159 | |
XM_008769345.1 | XP_008767567.1 | 4 - 5 | 2707 | 159 | ||
XM_008769346.1 | XP_008767568.1 | 5 - 6 | 2937 | 159 | ||
XM_008769347.1 | XP_008767569.1 | 5 - 6 | 1897 | 159 | ||
XM_008769348.1 | XP_008767570.1 | 5 - 6 | 2610 | 159 | ||
XM_008769349.1 | XP_008767571.1 | 5 - 6 | 2493 | 159 | ||
XM_008769350.1 | XP_008767572.1 | 4 - 5 | 2395 | 159 | ||
XM_008769351.1 | XP_008767573.1 | 3 - 4 | 1353 | 159 | ||
GenBank mRNA | BC088863.1 | - | 3 - 4 | 1474 | 159 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
