Assay Details
Gene Symbol: Pir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009474.1 | NP_001009474.1 | 8 - 9 | 832 | 120 | |
GenBank mRNA | BC088290.1 | - | 8 - 9 | 832 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P062462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114326 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P062462 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P062462 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P062462 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17743 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69078 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17883 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97740 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32132 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 95305 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 54129 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9150 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84563 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48476 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10937742 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937743 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937739 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937735 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937736 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937737 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937740 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937738 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937741 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937744 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10937734 | - | - |
Rat Genome 230 2.0 Array | 1377662_at | - | - |
Rat Genome 230 2.0 Array | 1377662_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377662_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1377662_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00010599.rn.1 | TC0X00001635.rn.1 | RTA TC |
Rat Expression Set 230 | 1377662_at | - | - |
Rat Expression Set 230 | 1377662_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937734 | - | - |
Rat Genome U34 Set | rc_AI176041_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21206296 | - | - |
ABI Rat Genome Survey Microarray | 21781591 | - | - |
ABI Rat Genome Survey Microarray | 22418811 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369072 | - | - |
RatRef-12 Expression BeadChip | ILMN_50170 | - | - |
