Assay Details
Gene Symbol: Cdh20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012748.1 | NP_001012766.1 | 2 - 3 | 538 | 70 | |
GenBank mRNA | AB121033.1 | - | 2 - 3 | 538 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
