Assay Details
Gene Symbol: Desi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013873.1 | NP_001013895.1 | 4 - 5 | 755 | 75 | |
XM_006250316.2 | XP_006250378.1 | 4 - 5 | 1105 | 75 | ||
XM_006250317.2 | XP_006250379.1 | 5 - 6 | 1519 | 75 | ||
XM_008769783.1 | XP_008768005.1 | 4 - 5 | 1062 | 75 | ||
XM_008769784.1 | XP_008768006.1 | 5 - 6 | 1238 | 75 | ||
GenBank mRNA | BC083584.1 | - | 4 - 5 | 755 | 75 | |
FQ226068.1 | - | 4 - 5 | 530 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
