Assay Details
Gene Symbol: Trim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014023.1 | NP_001014045.1 | 3 - 4 | 625 | 152 | |
XM_006229912.2 | XP_006229974.1 | 3 - 4 | 1111 | 152 | ||
GenBank mRNA | BC078926.1 | - | 3 - 4 | 625 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
