Assay Details
Gene Symbol: Trim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014023.1 | NP_001014045.1 | 4 - 5 | 855 | 128 | |
XM_006229912.2 | XP_006229974.1 | 4 - 5 | 1341 | 128 | ||
GenBank mRNA | BC078926.1 | - | 4 - 5 | 855 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
