Assay Details
Gene Symbol: Slco6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014270.1 | NP_001014292.1 | 1503 | 74 | ||
XM_006245580.2 | XP_006245642.1 | 1741 | 74 | |||
XM_006245583.2 | XP_006245645.1 | 1480 | 74 | |||
GenBank mRNA | BC083829.1 | - | 1503 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
