Assay Details
Gene Symbol: Spib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024286.1 | NP_001019457.1 | 3 - 4 | 135 | 106 | |
XM_006229111.2 | XP_006229173.1 | 2 - 3 | 884 | 106 | ||
XM_006229112.2 | XP_006229174.1 | 3 - 4 | 204 | 106 | ||
XM_006229113.2 | XP_006229175.1 | 2 - 3 | 884 | 106 | ||
XM_006229114.2 | XP_006229176.1 | 2 - 3 | 829 | 106 | ||
XM_006229115.2 | XP_006229177.1 | 3 - 4 | 149 | 106 | ||
XM_008759404.1 | XP_008757626.1 | 2 - 3 | 832 | 106 | ||
XM_008759405.1 | XP_008757627.1 | 2 - 3 | 185 | 106 | ||
GenBank mRNA | BC089881.1 | - | 3 - 4 | 135 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
