Assay Details
Gene Symbol: Bag5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008526.2 | NP_001008526.1 | 1823 | 137 | ||
XM_006240601.2 | XP_006240663.1 | 1983 | 137 | |||
XM_006240602.2 | XP_006240664.1 | 1923 | 137 | |||
XM_006240603.2 | XP_006240665.1 | 1899 | 137 | |||
GenBank mRNA | BC088418.1 | - | 1823 | 137 |
Gene Symbol: NEWGENE_1310847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764977.1 | XP_008763199.1 | 1 | 1712 | 137 | |
XM_008764978.1 | XP_008763200.1 | 2 | 1881 | 137 | ||
XM_008764979.1 | XP_008763201.1 | 2 | 1897 | 137 | ||
XM_008764980.1 | XP_008763202.1 | 2 | 1899 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
